Learn More
Sumatriptan Succinate, TRC
Product Code. 30136181
Change view
Click to view available options
Quantity:
10 mg
250 mg
500 mg
Unit Size:
10mg
250mg
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30136181 10 mg 10mg
30133663 500 mg 500mg
17971335 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30136181 Supplier Toronto Research Chemicals Supplier No. TRCS81000010MG
 Restricted Item
Estimated Shipment: 09-10-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 103628-48-4
Molecular Formula C14 H21 N3 O2 S . C4 H6 O4
Molecular Weight (g/mol) 413.49
Synonym Sumatriptan Succinate
IUPAC Name butanedioic acid;1-[3-[2-(dimethylamino)ethyl]-1H-indol-5-yl]-N-methylmethanesulfonamide
SMILES CNS(=O)(=O)Cc1ccc2[nH]cc(CCN(C)C)c2c1.OC(=O)CCC(=O)O
Specifications
Quantity 10 mg
InChI Formula InChI=1S/C14H21N3O2S.C4H6O4/c1-15-20(18,19)10-11-4-5-14-13(8-11)12(9-16-14)6-7-17(2)3;5-3(6)1-2-4(7)8/h4-5,8-9,15-16H,6-7,10H2,1-3H3;1-2H2,(H,5,6)(H,7,8)
Formula Weight 413.1621
Chemical Name or Material Sumatriptan Succinate