missing translation for 'onlineSavingsMsg'
Learn More

Sumatriptan Succinate EP Impurity E, Mikromol™

Product Code. 30051247
Change view
Click to view available options
Quantity:
25 mg
100 mg
4 x 25 mg
Unit Size:
100mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30051247 25 mg 25mg
17943616 100 mg 100mg
30459933 4 x 25 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30051247 Supplier Mikromol Supplier No. MM0310.050025

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Sumatriptan Succinate EP Impurity E
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (3-(2-Aminoethyl)-1H-indol-5-yl)-N-methylmethanesulfonamide
CAS 88919-22-6
Analyte or Component Names (3-(2-Aminoethyl)-1H-indol-5-yl)-N-methylmethanesulfonamide
Shipping Condition Room Temperature
Molecular Formula C12 H17 N3 O2 S
Certifications/Compliance ISO 9001
Synonym [3-(2-Aminoethyl)-1H-indol-5-yl]-N-methylmethanesulfonamide, Sumatriptan Succinate Imp. E (EP), Sumatriptan Imp. E (EP), [3-(2-Aminoethyl)-1H-indol-5-yl]-N-methylmethanesulfonamide, Sumatriptan amino, GR 32066X, Ph Eur Sumatriptan impurity E, BP Sumatriptan Impurity E, sumatriptan amino indole (GR 32066X), Ph Eur Sumatriptan Impurity Mixture, Sumatriptan Succinate Impurity E, Sumatriptan Impurity E
InChI Formula InChI=1S/C12H17N3O2S/c1-14-18(16,17)8-9-2-3-12-11(6-9)10(4-5-13)7-15-12/h2-3,6-7,14-15H,4-5,8,13H2,1H3
SMILES CNS(=O)(=O)Cc1ccc2[nH]cc(CCN)c2c1
IUPAC Name 1-[3-(2-aminoethyl)-1H-indol-5-yl]-N-methylmethanesulfonamide
Molecular Weight (g/mol) 267.35
Quantity 25 mg
Formula Weight 267.1041 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.