Learn More
Sumatriptan-d6, TRC
Specifications
Specifications
| Chemical Name or Material | Sumatriptan-d6 |
| CAS | 1020764-38-8 |
| Purity Grade Notes | HPLC |
| Molecular Formula | C14H15D6N3O2S |
| Quantity | 1 mg |
| Synonym | Sumatriptan D6 |
| InChI Formula | InChI=1S/C14H21N3O2S/c1-15-20(18,19)10-11-4-5-14-13(8-11)12(9-16-14)6-7-17(2)3/h4-5,8-9,15-16H,6-7,10H2,1-3H3/i2D3,3D3 |
| SMILES | [2H]C([2H])([2H])N(CCc1c[nH]c2ccc(CS(=O)(=O)NC)cc12)C([2H])([2H])[2H] |
| Molecular Weight (g/mol) | 301.44 |
| Formula Weight | 301.1731 |
| Show More |