missing translation for 'onlineSavingsMsg'
Learn More

Sulfur Set for ASTM D4045: S @ 0, 0.1, 0.5, 1.0, 2.5, 5.0, 10 μg/g in Isooctane

Product Code. 30326475
Change view
Click to view available options
Quantity:
Set (7)
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30326475 Set (7)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30326475 Supplier VHG Labs Supplier No. VHGSRCSET1A

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Sulfur
CAS 7704-34-9
Analyte or Component Names Sulfur
Shipping Condition Room Temperature
Molecular Formula S
Synonym Sulfur, Schwefel, Agri-Sul, Alphagran S 80, Asulfa-Supra, Tiger 90 J
InChI Formula InChI=1 S/S
Solution Type Mixture
SMILES [S]
Element S
IUPAC Name sulfur
Solvent or Matrix Isooctane
Industry Type Petrochemical Testing
Molecular Weight (g/mol) 32.065
Quantity Set (7)
Formula Weight 31.972
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.