Learn More
Witeg™ Sulfonation Flasks
Product Code. 11817800
Change view
Click to view available options
Capacity (Metric):
200 mL
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Unit Size
11817800 200 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11817800 Supplier Witeg™ Supplier No. 0 850 032
  • ONLINE EXCLUSIVE PRICE
    150.74 EUR
    127.60 EUR / Pack of 1
    Save 23.14 EUR
    15% Off
Estimated Shipment: 25-01-2027
An excellent vessel for use in pharmaceutical applications.
  • Available in either 100 or 200 mL capacities
  • With 4 side necks
  • Center neck socket ST 29/32
  • Side necks socket ST 19/26
Specifications
Capacity (Metric) 200 mL
Material Glass
For Use With (Application) Pharmaceutical
Neck Style 4 side necks, 1 center
Product Type Sulfonation flask
Socket Joint CN ST 29/32, SN ST 19/26