missing translation for 'onlineSavingsMsg'
Learn More

Sulfisomidine, TRC

Product Code. 30425386
Change view
Click to view available options
Quantity:
25 mg
50 mg
250 mg
Unit Size:
250mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30425386 250 mg 250mg
30421871 50 mg -
30423608 25 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30425386 Supplier Toronto Research Chemicals Supplier No. TRCS699378250MG
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 515-64-0
Molecular Formula C12 H14 N4 O2 S
Molecular Weight (g/mol) 278.33
Synonym Benzenesulfonamide, 4-amino-N-(2,6-dimethyl-4-pyrimidinyl)-, Sulfanilamide, N1-(2,6-dimethyl-4-pyrimidinyl)- (8CI), 2,4-Dimethyl-6-sulfanilamidopyrimidine, 2,6-Dimethyl-4-sulfanilamidopyrimidine, 4-Sulfanilamido-2,6-dimethylpyrimidine, 4-[[(2,6-Dimethylpyrimidin-4-yl)amino]sulfonyl]aniline, 6-(4-Aminobenzenesulfonamido)-2,4-dimethylpyrimidine, 6-(Sulfanilamido)-2,4-dimethyl-1,3-diazine, 6-(p-Aminobenzenesulfonamido)-2,4-dimethylpyrimidine, 6-(p-Aminophenylsulfonamido)-2,4-dimethylpyrimidine, 6-Sulfanilamido-2,4-dimethylpyrimidine, 6-[(p-Aminobenzenesulfonyl)amino]-2,4-dimethylpyrimidine, Aristamid, Aristogyn, Domain, Domian, Elcosin, Elcosine, Elkosil, Elkosin, Elkosine, Elkozin, Erycon, Mefenal, N1-(2,6-Dimethyl-4-pyrimidinyl)sulfanilamide, Sulfadimetine, Sulfaisodimerazine, Sulfamethine, Sulfasomidin, Sulfasomidine, Sulfisomidin, Sulfisomidine, Sulphasomidine, Sulphisomidine
IUPAC Name 4-amino-N-(2,6-dimethylpyrimidin-4-yl)benzenesulfonamide
SMILES Cc1cc(NS(=O)(=O)c2ccc(N)cc2)nc(C)n1

Specifications

Quantity 250 mg
InChI Formula InChI=1S/C12H14N4O2S/c1-8-7-12(15-9(2)14-8)16-19(17,18)11-5-3-10(13)4-6-11/h3-7H,13H2,1-2H3,(H,14,15,16)
Formula Weight 278.0837
Chemical Name or Material Sulfisomidine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.