Learn More
Sulfisomidine Sodium, TRC
Specifications
Specifications
| Chemical Name or Material | Sulfisomidine','Sodium |
| CAS | 2462-17-1 |
| Molecular Formula | C12 H13 N4 O2 S . Na |
| Quantity | 10 mg |
| Synonym | Benzenesulfonamide','4-amino-N-(2,6-dimethyl-4-pyrimidinyl)-','sodium salt (1:1), Benzenesulfonamide','4-amino-N-(2,6-dimethyl-4-pyrimidinyl)-','monosodium salt (9CI), Sodium','[N1-(2,6-dimethyl-4-pyrimidinyl)sulfanilamido]- (7CI), Sulfanilamide','N1-(2,6-dimethyl-4-pyrimidinyl)-','monosodium salt (8CI), Sulfanilamide','N1-(2,6-dimethyl-4-pyrimidinyl)-','sodium deriv. (6CI), Sodium sulfamethine, Sodium sulfasomidine, Sulfisomidine sodium |
| InChI Formula | InChI=1S/C12H13N4O2S.Na/c1-8-7-12(15-9(2)14-8)16-19(17,18)11-5-3-10(13)4-6-11;/h3-7H,13H2,1-2H3;/q-1;+1 |
| SMILES | [Na+].Cc1cc([N-]S(=O)(=O)c2ccc(N)cc2)nc(C)n1 |
| IUPAC Name | sodium;(4-aminophenyl)sulfonyl-(2,6-dimethylpyrimidin-4-yl)azanide |
| Molecular Weight (g/mol) | 300.31 |
| Formula Weight | 300.0657 |