missing translation for 'onlineSavingsMsg'
Learn More

Sulfisomidin, MedChemExpress

Product Code. 16418816
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16418816 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16418816 Supplier Medchem Express Supplier No. HYB1784
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Sulfisomidin (Sulfaisodimidine) is an orally active short-acting sulfonamide antibacterial. Sulfisomidin can be used for the research of lower urinary tract infections.

Sulfisomidin (Sulfaisodimidine) is an orally active short-acting sulfonamide antibacterial. Sulfisomidin can be used for the research of lower urinary tract infections.

Specifications

Chemical Name or Material Sulfisomidin
Physical Form Solid
CAS 515-64-0
Color White
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C12H14N4O2S
For Use With (Application) COVID-19-immunoregulation
Synonym Sulfaisodimidine
Solubility Information DMSO : ≥ 30 mg/mL (107.79 mM)
SMILES O=S(C1=CC=C(N)C=C1)(NC2=NC(C)=NC(C)=C2)=O
Molecular Weight (g/mol) 278.33
Formula Weight 278.33
Percent Purity 99.09%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.