Learn More
Tocris Bioscience™ Sulfasalazine
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
Description
Inhibitor of NF-κB activation. Inhibits in vitro growth of the human pancreatic cancer cell lines MIA PaCa-2 and PANC-1, and induces apoptosis in glioblastoma cell lines. Also inhibits the cystine-glutamate antiporter, system Xc (SXC). Anti-inflammatory. Induces ferroptosis.
Specifications
Specifications
| CAS | 599-79-1 |
| Quantity | 50 mg |
| Molecular Formula | C18H14N4O5S |
| Formula Weight | 398.39 |
| Purity | 97% |
| Molecular Weight (g/mol) | 398.393 |
| Chemical Name or Material | Sulfasalazine |