missing translation for 'onlineSavingsMsg'
Learn More

Sulfapyridine, LoGiCal

Product Code. 30459553
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30459553 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30459553 Supplier LoGiCal Supplier No. LGCFOR1596.00

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

LoGiCal delivers certified reference materials you can trust as neats, native solutions and stable isotope labelled solutions, with precise documentation and expert support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.

Sulfapyridine
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Sulfamethizole
CAS 144-82-1
Analyte or Component Names Sulfamethizole
Shipping Condition Room Temperature
Molecular Formula C9 H10 N4 O2 S2
Certifications/Compliance ISO 9001
Synonym Sulfasalazine Imp. J (EP), Sulfamethizole, 4-amino-N-(5-methyl-1,3,4-thiadiazol-2-yl)benzenesulfonamide, Benzenesulfonamide, 4-amino-N-(5-methyl-1,3,4-thiadiazol-2-yl)-, Sulfanilamide, N1-(5-methyl-1,3,4-thiadiazol-2-yl)- (6 CI,7 CI,8 CI), 4-Amino-N-(5-methyl-[1,3,4 ]thiadiazol-2-yl)benzenesulfonamide, 4-Amino-N-(5-methyl-[1,3,4 ]thiadiazol-2-yl)benzenesulfonamide, 4-Amino-N-(5-methyl-[1,3,4 ]thiadiazol-2-yl)benzenesulfonamide, 4-Amino-N-(5-methyl-[1,3,4 ]thiadiazol-2-yl)benzenesulfonamide, 4-Amino-N-(5-methyl-[1,3,4 ]thiadiazol-2-yl)benzenesulfonamide, 4-Amino-N-(5-methyl-[1,3,4 ]thiadiazol-2-yl)benzenesulfonamide, 4-Amino-N-(5-methyl-[1,3,4 ]thiadiazol-2-yl)benzenesulfonamide, 4-Amino-N-(5-methyl-[1,3,4 ]thiadiazol-2-yl)benzenesulfonamide, 4-Amino-N-(5-methyl-[1,3,4 ]thiadiazol-2-yl)benzenesulfonamide, 4-Amino-N-(5-methyl-[1,3,4 ]thiadiazol-2-yl)benzenesulfonamide, 4-Amino-N-(5-methyl-[1,3,4 ]thiadiazol-2-yl)benzenesulfonamide, 4-Amino-N-(5-methyl-[1,3,4 ]thiadiazol-2-yl)benzenesulfonamide, 4-Amino-N-(5-methyl-[1,3,4 ]thiadiazol-2-yl)benzenesulfonamide, 4-Amino-N-(5-methyl-[1,3,4 ]thiadiazol-2-yl)benzenesulfonamide, 4-Amino-N-(5-methyl-[1,3,4 ]thiadiazol-2-yl)benzenesulfonamide, 4-Amino-N-(5-methyl-[1,3,4 ]thiadiazol-2-yl)benzenesulfonamide, 4-Amino-N-(5-methyl-[1,3,4 ]thiadiazol-2-yl)benzenesulfonamide, 4-Amino-N-(5-methyl-[1,3,4 ]thiadiazol-2-yl)benzenesulfonamide, 4-Amino-N-(5-methyl-[1,3,4 ]thiadiazol-2-yl)benzenesulfonamide, 4-Amino-N-(5-methyl-[1,3,4 ]thiadiazol-2-yl)benzenesulfonamide, 4-Amino-N-(5-methyl-[1,3,4 ]thiadiazol-2-yl)benzenesulfonamide, 4-Amino-N-(5-methyl-[1,3,4 ]thiadiazol-2-yl)benzenesulfonamide, 4-Amino-N-(5-methyl-[1,3,4 ]thiadiazol-2-yl)benzenesulfonamide, 4-Amino-N-(5-methyl-[1,3,4 ]thiadiazol-2-yl)benzenesulfonamide, 4-Amino-N-(5-methyl-[1,3,4 ]thiadiazol-2-yl)benzenesulfonamide, 4-Amino-N-(5-methyl-[1,3,4 ]thiadiazol-2-yl)benzenesulfonamide, 4-Amino-N-(5-methyl-[1,3,4 ]thiadiazol-2-yl)benzenesulfonamide, 4-Amino-N-(5-methyl-[1,3,4 ]thiadiazol-2-yl)benzenesulfonamide, 4-Amino-N-(5-methyl-[1,3,4 ]thiadiazol-2-yl)benzenesulfonamide, 4-Amino-N-(5-methyl-[1,3,4 ]thiadiazol-2-yl)benzenesulfonamide, 4-Amino-N-(5-methyl-[1,3,4 ]thiadiazol-2-yl)benzenesulfonamide, 4-Amino-N-(5-methyl-[1,3,4 ]thiadiazol-2-yl)benzenesulfonamide
InChI Formula InChI=1 S/C9H10N4O2S2/c1-6-11-12-9(16-6)13-17(14,15)8-4-2-7(10)3-5-8/h2-5 H,10H2,1H3,(H,12,13)
SMILES Cc1nnc(NS(=O)(=O)c2ccc(N)cc2)s1
IUPAC Name 4-amino-N-(5-methyl-1,3,4-thiadiazol-2-yl)benzenesulfonamide
Industry Type Forensics
Molecular Weight (g/mol) 270.3313
Quantity 10 mg
Formula Weight 270.0245 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.