missing translation for 'onlineSavingsMsg'
Learn More

Sulfamethoxypyridazine, 99.04%, MP Biomedicals™

Product Code. 11465131 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
1 g
25 g
Unit Size:
1g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11465131 25 g 25g
11455131 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11465131 Supplier MP Biomedicals Supplier No. 0215671225
  • ONLINE EXCLUSIVE PRICE
    110.00 EUR
    103.95 EUR / 25g
    Save 6.05 EUR
    5% Off
 Restricted Item
Estimated Shipment: 25-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 80-35-3
Molecular Formula C11H12N4O3S
Molecular Weight (g/mol) 280.302
InChI Key VLYWMPOKSSWJAL-UHFFFAOYSA-N
Synonym sulfamethoxypyridazine, sulphamethoxypyridazine, sulfapyridazine, sulfalex, longin, midicel, sulfametoxipiridazine, spofadazine, sulfdurazin, 3-sulfa-6-methoxypyridazine
PubChem CID 5330
ChEBI CHEBI:102516
IUPAC Name 4-amino-N-(6-methoxypyridazin-3-yl)benzenesulfonamide
SMILES COC1=NN=C(C=C1)NS(=O)(=O)C2=CC=C(C=C2)N

Specifications

Quantity 25 g
Formula Weight 280.3
Percent Purity 0.9904
Physical Form Crystalline
Chemical Name or Material Sulfamethoxypyridazine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.