Learn More
Sulfamethizole, TRC
Product Code. 30415837
Change view
Click to view available options
Quantity:
1 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30415837 1 g
30418457 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30415837 Supplier Toronto Research Chemicals Supplier No. TRCS6890051G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 144-82-1
Molecular Formula C9 H10 N4 O2 S2
Molecular Weight (g/mol) 270.3313
Synonym Sulfamethizole, 4-amino-N-(5-methyl-1,3,4-thiadiazol-2-yl)benzenesulfonamide, Benzenesulfonamide, 4-amino-N-(5-methyl-1,3,4-thiadiazol-2-yl)-, Sulfanilamide, N1-(5-methyl-1,3,4-thiadiazol-2-yl)- (6CI,7CI,8CI), 2-Methyl-5-sulfanilamido-1,3,4-thiadiazole, 2-Sulfanilamido-5-methyl-1,3,4-thiadiazole, 4-Amino-N-(5-methyl-[1,3,4]thiadiazol-2-yl)benzenesulfonamide, 5-Methyl-2-sulfanilamido-1,3,4-thiadiazole, Ayerlucil, Famet, Lucosil, Methazol, Microsul, N-(5-Methyl-1,3,4-thiadiazol-2-yl)-4-aminobenzenesulfonamide, N1-(5-Methyl-1,3,4-thiadiazol-2-yl)sulfanilamide, Renasul, Rufol, Salimol, Sulfamethiazole, Sulfamethizol, Sulfamethizole, Sulfamethylthiadiazole, Sulfapyelon, Sulfstat, Sulfurine, Sulphamethizole, Tetracid, Thidicur, Thiosulfil, Thiosulfil Forte, Ultrasul, Urocydal, Urodiaton, Urolucosil, Urosulfin, VK 53
IUPAC Name 4-amino-N-(5-methyl-1,3,4-thiadiazol-2-yl)benzenesulfonamide
SMILES Cc1nnc(NS(=O)(=O)c2ccc(N)cc2)s1
Specifications
Quantity 1 g
InChI Formula InChI=1S/C9H10N4O2S2/c1-6-11-12-9(16-6)13-17(14,15)8-4-2-7(10)3-5-8/h2-5H,10H2,1H3,(H,12,13)
Formula Weight 270.0245
Percent Purity >95%
Chemical Name or Material Sulfamethizole
Grade HPLC
Product Title
Seleccione un problema
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.