Learn More
Sulfamethizole, Mikromol™
Description
Sulfamethizole
Specifications
Specifications
| Chemical Name or Material | Sulfamethizole |
| CAS | 144-82-1 |
| Analyte or Component Names | Sulfamethizole |
| Shipping Condition | Room Temperature |
| Molecular Formula | C9 H10 N4 O2 S2 |
| Certifications/Compliance | ISO 17025 |
| Synonym | Sulfasalazine Imp. J (EP), Sulfamethizole, 4-amino-N-(5-methyl-1,3,4-thiadiazol-2-yl)benzenesulfonamide, Benzenesulfonamide, 4-amino-N-(5-methyl-1,3,4-thiadiazol-2-yl)-, Sulfanilamide, N1-(5-methyl-1,3,4-thiadiazol-2-yl)- (6CI,7CI,8CI), 2-Methyl-5-sulfanilamido-1,3,4-thiadiazole, 2-Sulfanilamido-5-methyl-1,3,4-thiadiazole, 4-Amino-N-(5-methyl-[1,3,4]thiadiazol-2-yl)benzenesulfonamide, 5-Methyl-2-sulfanilamido-1,3,4-thiadiazole, Ayerlucil, Famet, Lucosil, Methazol, Microsul, N-(5-Methyl-1,3,4-thiadiazol-2-yl)-4-aminobenzenesulfonamide, N1-(5-Methyl-1,3,4-thiadiazol-2-yl)sulfanilamide, Renasul, Rufol, Salimol, Sulfamethiazole, Sulfamethizol, Sulfamethylthiadiazole, Sulfapyelon, Sulfstat, Sulfurine, Sulphamethizole, Tetracid, Thidicur, Thiosulfil, Thiosulfil Forte, Ultrasul, Urocydal, Urodiaton, Urolucosil, Urosulfin, VK 53, Sulfamethizol |
| InChI Formula | InChI=1S/C9H10N4O2S2/c1-6-11-12-9(16-6)13-17(14,15)8-4-2-7(10)3-5-8/h2-5H,10H2,1H3,(H,12,13) |
| SMILES | Cc1nnc(NS(=O)(=O)c2ccc(N)cc2)s1 |
| IUPAC Name | 4-amino-N-(5-methyl-1,3,4-thiadiazol-2-yl)benzenesulfonamide |
| Show More |