missing translation for 'onlineSavingsMsg'
Learn More

Sulfamethizole, MedChemExpress

Product Code. 16452106
Encompass_Preferred
Change view
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16452106 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16452106 Supplier Medchem Express Supplier No. HYB0333
 Restricted Item
Estimated Shipment: 17-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Sulfamethizole is a sulfathiazole antibacterial agent.

Sulfamethizole is a sulfathiazole antibacterial agent.

Specifications

Chemical Name or Material Sulfamethizole
Physical Form Solid
CAS 144-82-1
Color White
Quantity 500 mg
Purity Grade Notes Research
Molecular Formula C9H10N4O2S2
For Use With (Application) COVID-19-immunoregulation
Solubility Information DMSO : ≥ 100 mg/mL (369.92 mM) ∣H2O : < 0.1 mg/mL (insoluble)
SMILES O=S(C1=CC=C(N)C=C1)(NC2=NN=C(C)S2)=O
Molecular Weight (g/mol) 270.33
Formula Weight 270.33
Percent Purity 99.86%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.