Learn More
Sulfamethizole-d4 (benzene-d4), CDN
Specifications
Specifications
| Chemical Name or Material | Sulfamethizole phenyl-d4 |
| CAS | 2470130-12-0 |
| Molecular Formula | C9 2H4 H6 N4 O2 S2 |
| Quantity | 0.005 g |
| Synonym | Sulfamethizol D4 (benzene D4), 2-Methyl-5-sulfanilamido-1,3,4-thiadiazole-d4, 2-Sulfanilamido-5-methyl-1,3,4-thiadiazole-d4, 4-Amino-N-(5-methyl-[1,3,4 ]thiadiazol-2-yl)benzenesulfonamide-d4, 5-Methyl-2-sulfanilamido-1,3,4-thiadiazole-d4, Ayerlucil-d4, Famet-d4, Lucosil-d4, Methazol-d4, Microsul-d4, N-(5-Methyl-1,3,4-thiadiazol-2-yl)-4-aminobenzenesulfonamide-d4-d4, N1-(5-Methyl-1,3,4-thiadiazol-2-yl)sulfanilamide-d4, Renasul-d4, Rufol-d4, Salimol-d4, Sulfamethiazole-d4, Sulfamethizol-d4, Sulfamethylthiadiazole-d4, Sulfapyelon-d4, Sulfstat-d4, Sulfurine-d4, Sulphamethizole-d4, Tetracid-d4, Thidicur-d4, Thiosulfil-d4, Thiosulfil Forte-d4, Ultrasul-d4, Urocydal-d4, Urodiaton-d4, Urolucosil-d4, Urosulfin-d4, VK 53-d4, N1-(5-Methyl-1,3,4-thiadiazol-2-yl)sulfanilamide-d4, 4-Amino-N-(5-methyl-1,3,4-thiadiazol-2-yl)benzenesulfo, Sulfamethizol D4 (phenyl D4) |
| InChI Formula | InChI=1 S/C9H10N4O2S2/c1-6-11-12-9(16-6)13-17(14,15)8-4-2-7(10)3-5-8/h2-5 H,10H2,1H3,(H,12,13)/i2D,3 D,4 D,5 D |
| SMILES | [2 H]c1c([2 H])c(c([2 H])c([2 H])c1N)S(=O)(=O)Nc2nnc(C)s2 |
| IUPAC Name | 4-amino-2,3,5,6-tetradeuterio-N-(5-methyl-1,3,4-thiadiazol-2-yl)benzenesulfonamide |
| Molecular Weight (g/mol) | 274.36 |
| Formula Weight | 274.0496 g/mol |
| Show More |