missing translation for 'onlineSavingsMsg'
Learn More

Sulfamethazine sodium salt, MP Biomedicals™

Product Code. 11495121 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
100 g
250 g
Unit Size:
100g
250g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11495121 250 g 250g
11485121 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11495121 Supplier MP Biomedicals Supplier No. 0215670983

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 1981-58-4
Molecular Formula C12H13N4NaO2S
Molecular Weight (g/mol) 300.312
InChI Key NGIVTUVVBWOTNT-UHFFFAOYSA-N
Synonym sulfamethazine sodium salt, sulfamethazine sodium, sulfadimidine sodium, sodium sulfamethazine, sodium sulfametazine, sodium sulfamethamide, sodium sulfamezathine, bovibol, vesadin, sodium sulfamethiazine
PubChem CID 13456556
IUPAC Name sodium;(4-aminophenyl)sulfonyl-(4,6-dimethylpyrimidin-2-yl)azanide
SMILES CC1=CC(=NC(=N1)[N-]S(=O)(=O)C2=CC=C(C=C2)N)C.[Na+]

Specifications

Quantity 250 g
Formula Weight 300.3
Physical Form Solid
Chemical Name or Material Sulfamethazine sodium salt
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.