missing translation for 'onlineSavingsMsg'
Learn More

Sulfameter, MedChemExpress

Product Code. 16448176
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16448176 100 mg 100mg
16422066 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16448176 Supplier Medchem Express Supplier No. HYB0213
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

MedChemExpress Sulfameter (Sulfametoxydiazine; 5-Methoxysulfadiazine) is an effective long-acting sulfonamide antibiotic with antibacterial activities. Sulfameter can be used for the research of urinary tract infections and lepriasis.

Sulfameter (Sulfametoxydiazine

Specifications

Chemical Name or Material Sulfameter
Physical Form Solid
CAS 651-06-9
Color White
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C11H12N4O3S
For Use With (Application) COVID-19-immunoregulation
Synonym Sulfametoxydiazine 5-Methoxysulfadiazine
Solubility Information DMSO : ≥ 100 mg/mL (356.76 mM) ∣H2O : 0.67 mg/mL (2.39 mM; Need ultrasonic)
SMILES O=S(C1=CC=C(N)C=C1)(NC2=NC=C(OC)C=N2)=O
Molecular Weight (g/mol) 280.3
Formula Weight 280.3
Percent Purity 99.89%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.