Learn More
Sulfameter, Dr. Ehrenstorfer
Product Code. 30310807
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30310807 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30310807 Supplier Dr. Ehrenstorfer Supplier No. DREC16995150100MG
 Restricted Item
Estimated Shipment: 17-08-2026
Add to basket
Add to basket
Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Sulfameter
CAS 651-06-9
Analyte or Component Names Sulfameter
Shipping Condition +20°C
Molecular Formula C11 H12 N4 O3 S
Certifications/Compliance ISO 17034
Synonym Sulfameter
InChI Formula InChI=1S/C11H12N4O3S/c1-18-9-6-13-11(14-7-9)15-19(16,17)10-4-2-8(12)3-5-10/h2-7H,12H2,1H3,(H,13,14,15)
SMILES COc1cnc(NS(=O)(=O)c2ccc(N)cc2)nc1
IUPAC Name 4-amino-N-(5-methoxypyrimidin-2-yl)benzenesulfonamide
Molecular Weight (g/mol) 280.3
Quantity 100 mg
Formula Weight 280.063 g/mol