Learn More
Sulfacetamide, TRC
Product Code. 30416059
Change view
Click to view available options
Quantity:
1 g
10 g
50 g
Unit Size:
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30416059 10 g -
30417386 50 g 50g
30418563 1 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30416059 Supplier Toronto Research Chemicals Supplier No. TRCS68896010G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 144-80-9
Molecular Formula C8 H10 N2 O3 S
Molecular Weight (g/mol) 214.24
Synonym N-[(4-Aminophenyl)sulfonyl]acetamide, Sulfacetamide, Sulfadimidine Imp. E (Pharmeuropa)
IUPAC Name N-(4-aminophenyl)sulfonylacetamide
SMILES CC(=O)NS(=O)(=O)c1ccc(N)cc1
Specifications
Quantity 10 g
InChI Formula InChI=1S/C8H10N2O3S/c1-6(11)10-14(12,13)8-4-2-7(9)3-5-8/h2-5H,9H2,1H3,(H,10,11)
Formula Weight 214.0412
Chemical Name or Material Sulfacetamide