accessibility menu, dialog, popup

UserName

CAS RN 144-80-9

CH3ONHSOONH2

CAS RN 144-80-9

IUPAC Name: N-(4-aminobenzenesulfonyl)acetamide
Synonyms: sulfacetamide op-sulfa 30 sulfacetamide (substance) sulf-15 sulf-10 bleph-10 liquifilm monosodium salt, sulfacetamide N-acetylsulfanilamine sulten-10 sulfacetamide sodium usp
Molecular Weight (g/mol): 214.24
Molecular Formula: C8H10N2O3S
InChi Key: SKIVFJLNDNKQPD-UHFFFAOYSA-N
SMILES: CC(=O)NS(=O)(=O)C1=CC=C(N)C=C1
Molecular Weight (g/mol) 
  • (2)
  • (6)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Certifications/Compliance 
  • (1)
  • (2)
  • (1)
Percent Purity 
  • (2)
Type 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results