Learn More
Sucrose-1,1,6,6,6’,6’-d6 (Major), TRC
Specifications
Specifications
| Chemical Name or Material | Sucrose-1,1,6,6,6',6'-d6 |
| CAS | 955943-30-3 |
| Molecular Formula | C12H16D6O11 |
| Quantity | 50 mg |
| Synonym | β-D-Fructofuranosyl α-D-Glucopyranoside-d6, (+)-Sucrose-d6, Amerfond-d6, Beet Sugar-d6, Cane Sugar-d6, Carrare C 10-d6, Compressuc-d6, Confectioner's Sugar-d6, D-(+)-Saccharose-d6, D-(+)-Sucrose-d6, D-Sucrose-d6, Frost Sugar-d6, GNE 410-d6, Granulated Sugar-d6, Khandsari-d6, Manalox AS-d6, Microse-d6, NSC 406942-d6, Rock Candy-d6, Saccharose-d6, Saccharum-d6, Sucralox-d6, Sucrose NF-d6, Sugar-d6, White Sugar-d6, α-D-Glucosyl-β-D-fructofuranoside-d6 |
| InChI Formula | InChI=1S/C12H22O11/c13-1-4-6(16)8(18)9(19)11(21-4)23-12(3-15)10(20)7(17)5(2-14)22-12/h4-11,13-20H,1-3H2/t4-,5-,6-,7-,8+,9-,10+,11?,12+/m1/s1/i1D2,2D2,3D2 |
| SMILES | [2H]C([2H])(O)[C@H]1OC(O[C@@]2(O[C@@H]([C@@H](O)[C@@H]2O)C([2H])([2H])O)C([2H])([2H])O)[C@H](O)[C@@H](O)[C@@H]1O |
| Molecular Weight (g/mol) | 348.33 |
| Formula Weight | 348.1539 |