Learn More
Succinic Anhydride, Sigma-Aldrich™
Product Code. 15876250
Change view
Click to view available options
Quantity:
500 g
1 kg
50 kg
Unit Size:
1kg
500g
50kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15876250 1 kg 1kg
15866250 500 g 500g
15838380 50 kg 50kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 15876250 Supplier Sigma Aldrich Supplier No. 8006831000
 Restricted Item
Estimated Shipment: 20-11-2026
For synthesis
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 108-30-5
Specifications
Melting Point 116°C to 120°C
Density 1.23 g/cm3 at 20°C
Boiling Point 261°C/1013 hPa
MDL Number MFCD00005525
Quantity 1 kg
Synonym Succinic anhydride, Succinic anhydride
Molecular Weight (g/mol) 100.07
Grade Research