missing translation for 'onlineSavingsMsg'
Learn More

Succinic Acid, Sigma-Aldrich™

Product Code. 15876720
Change view
Click to view available options
Quantity:
250 g
1 kg
Unit Size:
1kg
250g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15876720 250 g 250g
15891731 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15876720 Supplier Sigma Aldrich Supplier No. 8222600250
 Restricted Item
Estimated Shipment: 20-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For synthesis

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 110-15-6

Specifications

Melting Point 184°C to 186°C (lit.)
Boiling Point 235°C/1013 hPa,235°C
Density 1.56 g/cm3 at 20°C
Quantity 250 g
Molecular Formula HOOCCH2CH2COOH
MDL Number MFCD00002789
Synonym Butanedioic acid, Butanedioic acid, Butanedioic acid
Molecular Weight (g/mol) 118.09
Grade Ungraded
Chemical Name or Material Succinic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.