Learn More
Suberyl Glycine-d4, TRC
Specifications
Specifications
| Chemical Name or Material | Suberyl Glycine-d4 |
| CAS | 2469259-73-0 |
| Molecular Formula | C10 D4 H13 N O5 |
| Quantity | 1 mg |
| Synonym | 8-[(Carboxymethyl)amino]-8-oxooctanoic-2,2,7,7-d4 acid, Suberylglycine-D4 (suberyl-2,2,7,7-D4), Suberyl glycine-d4, N-(Suberyl-2,2,7,7-d4)glycine, N-Suberyl-2,2,7,7-d4-glycine |
| InChI Formula | InChI=1S/C10H17NO5/c12-8(11-7-10(15)16)5-3-1-2-4-6-9(13)14/h1-7H2,(H,11,12)(H,13,14)(H,15,16)/i5D2,6D2 |
| SMILES | [2H]C([2H])(CCCCC([2H])([2H])C(=O)NCC(=O)O)C(=O)O |
| IUPAC Name | 8-(carboxymethylamino)-2,2,7,7-tetradeuterio-8-oxooctanoic acid |
| Molecular Weight (g/mol) | 235.27 |
| Formula Weight | 235.136 |