Learn More
Cole-Parmer™ Digital Colony Counter Optional Magnifier
Product Code. 11446508
Change view
Click to view available options
Magnification:
1.7x
3x
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Magnification Unit Size
11446508 1.7x Each
12088183 3x Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11446508 Supplier Cole-Parmer™ Supplier No. 1421260
  • ONLINE EXCLUSIVE PRICE
    153.00 EUR
    110.63 EUR / Pack of 1
    Save 42.37 EUR
    27% Off
Estimated Shipment: 11-12-2026
Use this magnifier with CC-200 Colony Counters.
Use this magnifier with Stuart™ SC6 Colony Counters.
  • 1.7x and 3x magnifier for CC-200 Colony Counter.
  • Pressure sensitive counting.
  • Average count facility.
  • Bright white energy saving LED lighting.
  • With BioCote antimicrobial protection.
  • Audible count confirmation.
  • Choice of light or dark background.
  • Connectivity to printer or computer.
Specifications
For Use With (Equipment) The Colony Counter Stuart SCPPLUS
Includes BioCote Antimicrobial protection
Quantity 1 Ea.
Magnification 1.7x
Product Type Lens, Magnifying