Learn More
Cole-Parmer™ WS-100 Series Stuart™ Water Still
Product Code. 13145990
Change view
Click to view available options
Voltage:
220 V
240 V
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Voltage Unit Size
13145990 240 V Each
12068990 220 V Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 13145990 Supplier Cole-Parmer™ Supplier No. 9929300
  • ONLINE EXCLUSIVE PRICE
    1850.00 EUR
    1465.58 EUR / Pack of 1
    Save 384.42 EUR
    20% Off
View all promotions
 Restricted Item
Estimated Shipment: 26-11-2026
Save money while maintaining high performance with this WS-100 Series Stuart Water Still.
  • Two independent thermostats prevent overheating.
  • Features a sturdy metal stand for either wall or bench mounting.
  • Operate directly from tap water.
  • This basic economical water still is perfect for general laboratory and educational distillations.
  • Borosilicate glass boiler and condenser provide the highest purity distillate.
  • The unique condenser design ensures that the distilled water droplets remain in contact with the cooling coil for an extended period for maximum energy transfer and immediate cold distilled water.
  • Condenser with screw thread connections and plastic 3/8 in. ID hose barb fittings allow quick and safe tubing change without worrying about breaking the glassware.
  • Easily descale without dismantling the glassware using the built-in acid addition funnel and wastewater drain.
  • Long-life chromium-plated metal heater is protected by twin safety thermostats.
Specifications
Product Type Merit Water Still
Output Flow Rate 4 L/hr.
Electrical Requirements 220/240 V 50/60 Hz 3 kW
Frequency 50/60 Hz
Phase 1
Voltage 240 V
Inlet Water Temperature (Metric) 25°C to 35°C
Model W4000
Depth (English) 5.9 in.
Depth (Metric) 150 mm
Height (English) 17.7 in.
Height (Metric) 450 mm
Width (English) 19.7 in.
Width (Metric) 500 mm