Learn More
IBA Lifesciences Strep-Tactin™XT 4Flow™ High Capacity Resin
Product Code. p-7230525 Shop All IBA Lifesciences Products
Click to view available options
Quantity:
4 mL
20 mL
50 mL
1000 mL
Unit Size:
1 set
1000mL
Product Code. 16674724
Brand: IBA Lifesciences 25030002
Strep-Tactin™XT 4Flow™ high capacity resin is a 50% suspension to purify Strep-tag™II or Twin-Strep-tag™ fusion proteins
  • Strep-Tactin™XT 4Flow™ high capacity is a 4% agarose coupled with the streptavidin variant Strep-Tactin™XT and applicable for purification of Strep-tag™II or Twin-Strep-tag™ fusion proteins.
  • Due to the broad pH compatibility, excellent pressure stability, outstanding target purity (> 95%), and highest binding affinity, the resin is recommend for all protein classes, especially for low abundant or challenging proteins.
  • The low concentrated agarose enables the simple attachment of larger proteins to the stationary phase, making it particularly suitable for purification of high-molecular-weight targets.
  • In addition, Strep-Tactin™XT 4Flow™ high capacity allows the application of denaturing agents such as urea.
  • In comparison to Strep-Tactin™XT 4Flow™, Strep-Tactin™XT 4Flow™ high capacity has a higher density of immobilized Strep-Tactin™XT, which leads to a 2-fold higher yield of purified protein.
  • The suspension can be used for packing of gravity flow columns since the resin is optimized for column affinity chromatography, rather than for batch purification.
  • To pack a column with 1 ml resin bed volume, 2 ml 50% suspension are necessary.
Specifications
Matrix 4% agarose, crosslinked
Particle Size 50 to 150 μm
Quantity 4 mL
Target Twin-Strep-Tag™ and Strep-tag™II
Format Pre-packed Gravity Flow Column
For Research Use Only.