Learn More
Lenz Laborglasintrumente Straight Tubing Connectors
Product Code. 10030311
Change view
Click to view available options
Diameter (Metric) Outer:
8 mm
9 mm
Unit Size:
Pack of 10
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) Outer unitSize
10030311 9 mm Pack of 10
10439172 8 mm Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 10030311 Supplier Lenz Laborglasintrumente Supplier No. 01600009
  • ONLINE EXCLUSIVE PRICE
    12.00 EUR
    8.89 EUR / Pack of 10
    Save 3.11 EUR
    25% Off
Estimated Shipment: 18-08-2026
Add to basket
Add to basket
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
  • Tubing connectors
  • Straight
  • With two glass olives
  • Made of DURAN™ tubing
Specifications
Material Borosilicate Glass (3.3)
Diameter (Metric) Outer 9 mm
Overall Length (Metric) 115 NS
Includes Two glass olives