Learn More
Thermo Scientific™ Store Image and Database Management Software, enterprise edition
Product Code. 17139625 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
1 each
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17139625 1 each Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 17139625 Supplier Thermo Scientific™ Supplier No. S0100015
 Restricted Item
Estimated Shipment: 26-11-2026
Used for managing the images and data automatically transferred from a Thermo Scientific™ High Content Product
  • Thermo Scientific™ Store Image and Database Management Software, Enterprise Edition, is used for managing the images and data automatically transferred from a Thermo Scientific™ High Content Product.
  • It allows for rapid, highly scalable, secure, storage of images and data with the ability to query.
  • Features of the Store Image and Database Management Software, Enterprise Edition:
  • Automatic Transfer of Images and Data—Centralized data storage allowing data and image access from multiple locations
  • Accessible and Linked —Data is seamlessly linked to its associated image
  • Visualize—View images and data linked together and easily compare images
  • export—Built in export formats for software such as Spotfire™, Excel™, De Novo™ Software, and GeneData Screener™ programs
  • The Store Image and Database Management Software maintains data and images on a SQL™ or Oracle™ database.
TRUSTED_SUSTAINABILITY
Specifications
Product Type Image Management, Database Management
For Use With (Application) Data/Image Management
For Use With (Equipment) HCA/HCS
License Enterprise
Operating System CellInsight
Quantity 1 each