missing translation for 'onlineSavingsMsg'
Learn More

STO-609, MedChemExpress

Product Code. 16490378
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16490378 5 mg 5mg
16473857 50 mg 50mg
16463696 25 mg 25mg
16447918 10 mg 10mg
16427726 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16490378 Supplier Medchem Express Supplier No. HY19805

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress STO-609 is a selective and cell-permeable inhibitor of the Ca2+/calmodulin-dependent protein kinase kinase (CaM-KK), with Ki values of 80 and 15 ng/mL for recombinant CaM-KKα and CaM-KKβ, respectively. STO-609 inhibits AMP-activated protein kinase kinase (AMPKK) activity in HeLa cell lysates with an IC50 ∼0.02 g/ml.

STO-609 is a selective and cell-permeable inhibitor of the Ca2+/calmodulin-dependent protein kinase kinase (CaM-KK), with Ki values of 80 and 15 ng/mL for recombinant CaM-KKα and CaM-KKβ, respectively. STO-609 inhibits AMP-activated protein kinase kinase (AMPKK) activity in HeLa cell lysates with an IC50 ∼0.02 g/ml.

Specifications

Chemical Name or Material STO-609
CAS 52029-86-4
Color Yellow
Purity Grade Notes Research
Molecular Formula C19H10N2O3
Quantity 5 mg
Solubility Information DMSO : 6.25 mg/mL (19.89 mM; ultrasonic and warming and heat to 80°C) ∣DMF : 5.56 mg/mL (17.69 mM; Need ultrasonic) ∣H2O : < 0.1 mg/mL (insoluble)
SMILES O=C(C1=C2C3=C(C4=NC5=CC=CC=C5N4C(C3=CC=C2)=O)C=C1)O
Molecular Weight (g/mol) 314.29
Formula Weight 314.29
Percent Purity 98.0%
Grade Research
Physical Form Powder
For Use With (Application) Metabolism-sugar/lipid metabolism
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.