Learn More
Lovibond™ Stirring rod
Product Code. 11367320
Change view
Click to view available options
Type:
Stirring Rod
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Type Unit Size
11367320 Stirring Rod Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11367320 Supplier Lovibond™ Supplier No. 364120
  • ONLINE EXCLUSIVE PRICE
    31.30 EUR
    26.64 EUR / Pack of 1
    Save 4.66 EUR
    14% Off
Estimated Shipment: 17-12-2026
Plastic Stirring rod
Stirring rod, 13 cm length, set of 10.
Specifications
Type Stirring Rod
Length (Metric) 13 cm