Specifications
| Color | White |
| For Use With (Equipment) | Hotplate-Stirrer e-G51HS07C 120V US, Hotplate-Stirrer e-G51HSRDM 120V US, Hotplate-Stirrer e-G51HS10C 120V US, Stirrer e-G51ST07C 120V US, Hotplate-Stirrer e-G71HSRDM 120V US, Hotplate-Stirrer e-G71HS10C 120V US, Hotplate-Stirrer e-G71HS07C 120V US |
| Type | Stir Bar Retriever |
| Material | Plastic |