missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
STING/TMEM173 Monoclonal antibody specifically detects STING/TMEM173 in Human samples. It is validated for Western Blot, ELISA
Specifications
Specifications
| Antigen | STING/TMEM173 |
| Applications | Western Blot, ELISA |
| Classification | Monoclonal |
| Clone | 7C7 |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:500-1:5000, ELISA |
| Formulation | PBS, pH 7.4, 150mM NaCl and 50% glycerol |
| Gene Alias | endoplasmic reticulum IFN stimulator, Endoplasmic reticulum interferon stimulator, ERIS, FLJ38577, hMITA, hSTING, Mediator of IRF3 activation, MITA, mitochondrial mediator of IRF3 activation, MPYS, NET23, N-terminal methionine-proline-tyrosine-serine plasma membrane tetraspanner, SAVI, Stimulator of interferon genes protein, stimulator of interferon protein, sting, STING-beta, TMEM173, transmembrane protein 173 |
| Host Species | Rabbit |
| Immunogen | A synthesized peptide derived from Human STING/TMEM173 [UniProt Q86WV6] |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?