Learn More
Sampling Systems™ SteriWare™ Sterile Disposable LiquiThief
BIOPHARMA
Product Code. 15724758 Shop All Sampling Systems Products
Change view
Click to view available options
Capacity (Metric):
290 mL
Unit Size:
Pack of 20
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) unitSize
15724758 290 mL Pack of 20
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15724758 Supplier Sampling Systems™ Supplier No. 8050H1500S
Only null left
Add to basket
Add to basket
Sampling Systems™ SteriWare™ Sterile Disposable LiquiThief ideal for use with low viscocity liquids.
  • Assembled and packed in a Class 100,000 Cleanroom
  • Individually bagged
  • Made from FDA, EC 1935/2004 & EU 10/2011 conforming materials
  • BSE/TSE free
  • Full batch traceability
  • Available pre-sterilized (SAL 10-6)
  • At the required depth pull up the handle
  • The further the handle is pulled, the larger the sample volume
  • Push handle down to expel the sample
  • No cross contamination
  • No cleaning required
Specifications
Capacity (Metric) 290 mL
Diameter (Metric) Outside 21 mm
Description Sterile, Individually bagged
Length (Metric) 1500 mm
No. per Case 20