Description
- Moulded and packed in a Class 100,000 Cleanroom
- Individually bagged
- Made from FDA, EC 1935/2004 and EU 10/2011 conforming material
- BSE/TSE free
- Graduation marks in mL
- Full batch traceability
- Available pre-sterilized (SAL 10-6)
Specifications
Specifications
| Capacity (Metric) | 500 mL |
| Material | Polypropylene |
| Quantity | 35 Non-Sterile |
| Sterility | Non-sterile |
| Graduated | Yes |