Learn More
Buerkle™ SteriPlast™ Blue Scraper
BIOPHARMA
Product Code. 15644134 Shop All Bürkle Products
Change view
Click to view available options
Dimensions (L x W):
200 x 80 mm (7.8 x 3.1 in.)
Unit Size:
Pack of 10
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Dimensions (L x W) Unit Size
15644134 200 x 80 mm (7.8 x 3.1 in.) Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15644134 Supplier Buerkle™ Supplier No. 53783030
Estimated Shipment: 10-12-2026
For scraping off, smoothing or cleaning and removing residue, sterilized by gamma rays
  • Individually packaged and sterilised by gamma rays
  • Production, assembly and packaging according to clean room class 7
  • Can be used as part of HACCP/IFS/BRC foreign object management
Compliance
EU food requirements
TRUSTED_SUSTAINABILITY
Specifications
Product Type Scraper
Sterility Sterile
Color Blue
Dimensions (L x W) 200 x 80 mm (7.8 x 3.1 in.)
Disposable Single-use
For Use With (Application) Scraping Off, Smoothing or Cleaning and Removing Residue
Material Polystyrene