Learn More
Thermo Scientific™ Sterilin™ Special Petri Dishes
Description
Perform nonstandard applications with Thermo Scientific™ Sterilin™ Special Petri Dishes. Minimum order quantities apply.
- Aseptically manufactured
- In vitro use only
Ordering Alerts:
Not available in all geographic regions
Specifications
Specifications
| Certifications/Compliance | 90mm dishes tested in accordance with EN ISO 24998:2008 standard |
| Color | Clear |
| Shape | Round |
| Sterility | Aseptically Manufactured |
| Type | Petri Dish |
| Well Count | 2 |
| No. per Case | 500 |
| No. per Pack | 20 |
| Vent | 3 |
| Height (Metric) | 16 mm |
| Show More |