Learn More
Fisherbrand™ Sterile Sample Bags
Product Code. 17953182 Shop All Fisherbrand Traceable Products
Change view
Click to view available options
Dimensions (L x W):
4.5 x 9 in.
12 x 18 in.
15 x 20 in.
Quantity:
100/Pk.
250/Pk.
500/Pk.
Unit Size:
Pack of 100
Pack of 250
Pack of 500
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Dimensions (L x W) Quantity Unit Size
17953182 12 x 18 in. 250/Pk. Pack of 250
17943182 4.5 x 9 in. 500/Pk. Pack of 500
17903192 15 x 20 in. 100/Pk. Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 17953182
Save Up to 60% on Liquid Handling
Use promo code 28028
View all promotions
Estimated Shipment: 07-12-2026
An economical and efficient way to collect, contain and carry samples.
  • These sample bags are ideal for transportation and storage of solids, semisolids, and liquids.
  • Sterile
  • Highly resistant
  • Wire closure for a superior seal
Specifications
Autoclavable Not Autoclavable
Color Clear
Material Polyethylene
Sterility Sterile
Quantity 250/Pk.
Dimensions (L x W) 12 x 18 in.
Height (Metric) 457 mm
Length (Metric) 305 mm
Thickness (Metric) 100μm