Learn More
Corning™ Sterile Polystyrene Inoculating Loops
Product Code. 12682275
Change view
Click to view available options
Volume:
1 μL
10 μL
Product Type:
Inoculating Loop
Inoculating Loop, Needle Tip
Inoculating Loop, Spherical Tip
Unit Size:
Pack of 3000
Pack of 9000
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume Product Type Unit Size
12682275 1 μL Inoculating Loop, Needle Tip Pack of 9000
11733124 1 μL Inoculating Loop, Needle Tip Pack of 3000
15241065 1 μL Inoculating Loop, Spherical Tip Pack of 9000
11763124 10 μL Inoculating Loop, Needle Tip Pack of 3000
11769515 10 μL Inoculating Loop Pack of 9000
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 12682275 Supplier Corning™ Supplier No. OS101
  • ONLINE EXCLUSIVE PRICE
    535.00 EUR
    525.26 EUR / Pack of 9000
    Save 9.74 EUR
    1% Off
Estimated Shipment: 18-03-2027
Constructed of flexible polystyrene for optimum retrieval of an inoculum from a culture. Gosselin™ Sterile Polystyrene Inoculating Loops provide a superior solution for transferring microscopic organisms, inoculum, or streaking.
  • Polystyrene (PS) inoculating loops
  • Spherical or needle extremity
  • 1 μl or 10 μl
TRUSTED_SUSTAINABILITY
Specifications
Volume 1 μL
Length (Metric) 192 mm
Material Polystyrene
Sterility Sterile
Color Natural
Product Type Inoculating Loop, Needle Tip
Packaging Type Tamper evident zip bag