Learn More
Brand™ Polystyrene Cylindrical Test Tubes
Product Code. 10448692 Shop All Brand Products
Change view
Click to view available options
Diameter (Metric):
16 mm
Unit Size:
Pack of 2000
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) Unit Size
10448692 16 mm Pack of 2000
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10448692 Supplier Brand™ Supplier No. 114715
  • ONLINE EXCLUSIVE PRICE
    149.00 EUR
    110.20 EUR / Pack of 2000
    Save 38.80 EUR
    26% Off
Estimated Shipment: 16-12-2026
Manufactured from transparent. BRAND™ Polystyrene Cylindrical Test Tubes have an RCF max of 3000.
  • Universal
  • RCF Max: 3000
  • Manufactured from transparent polystyrene
TRUSTED_SUSTAINABILITY
Specifications
Color Transparent
Description 3000
Length (Metric) 100 mm
Material Polystyrene
Packaging 2000Pack
Sterility Non-sterile
Diameter (Metric) 16 mm