Learn More
Fisherbrand™ Sterile Polyethylene Sampling Bags
Product Code. 11532773 Shop All Fisherbrand Traceable Products
Change view
Click to view available options
Capacity (Metric):
540 mL
720 mL
2250 mL
3500 mL
Includes:
No Writing Area
Writing Area
Unit Size:
Pack of 1000
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Includes Unit Size
11532773 540 mL No Writing Area Pack of 1000
11562783 2250 mL Writing Area Pack of 1000
11572783 3500 mL Writing Area Pack of 1000
11532783 540 mL Writing Area Pack of 1000
11542783 720 mL Writing Area Pack of 1000
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 11532773
Sample Preparation and Storage Essentials
Use promo code 27996
View all promotions
Estimated Shipment: 16-11-2026
Add to basket
Add to basket
Designed with a twirl round metal wire closure system and puncture proof safety tabs. Fisherbrand™ Sterile Polyethylene Sampling Bags are leakproof when sealed.
  • Manufactured with pliant, highly resistant virgin polyethylene 
  • Sterile transparent polyethylene bags without side seals
  • Choose clear or white marking area for written notations
  • Convenient pull tabs to open bag without touching the edges
  • Ideal for use with solid, semi-solid or liquid samples
  • For use in all laboratories where samples need to be collected into a sterile environment.
Specifications
Color Clear
For Use With (Application) Use in all laboratories where samples need to be collected into a sterile environment
Includes No Writing Area
Material Polyethylene
Sterility Sterile
Quantity 1000 Pk.
Dimensions (L x W) 229 x 114 mm
Capacity (Metric) 540 mL
Thickness (Metric) 63 μm