Learn More
Greiner Bio-One Sterile Inoculation Loop
Product Code. 10707682
Change view
Click to view available options
Color:
Blue
White
Yellow
Quantity:
2000/Pk.
600/Pk.
Unit Size:
Pack of 2000
Pack of 600
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Quantity Unit Size
10707682 Blue 600/Pk. Pack of 600
10659021 White 2000/Pk. Pack of 2000
10342651 Yellow 600/Pk. Pack of 600
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 10707682 Supplier Greiner Bio-One Supplier No. 731171
  • ONLINE EXCLUSIVE PRICE
    106.00 EUR
    96.71 EUR / Pack of 600
    Save 9.29 EUR
    8% Off
Estimated Shipment: 08-12-2026
Transfers inoculum for streaking. Greiner Bio-One™ Sterile Inoculation Loops are ideal for retrieving an inoculum from a culture of microorganisms.
  • Flexible loops for easier collection and inoculation
  • Sterilized by irradiation
  • For research only; not for use in diagnostic procedures, unless otherwise indicated
TRUSTED_SUSTAINABILITY
Specifications
Disposable Single-use
Volume 10 μL
Length (English) 7.87 in.
Length (Metric) 200 mm
Material Polystyrene
Sterility Sterile
Size Standard
Color Blue
Product Type Inoculating Loop
Quantity 600/Pk.