Learn More
DWK Life Sciences WHEATON™ CompletePAK™ Sterile Components Seals
Shop All Wheaton Science Products ProductsDescription
- Sterile
- Flip-off Seal
Certifications
USP <788>, <85> and <71>; Container Closure Integrity (CCI) certified
Specifications
Specifications
| Color (Seal) | Red |
| Material | Aluminum |
| Sterility | Sterile |
| No. per Case | 230 |
| Product Line | Wheaton CompletePAK |
| Closure Type | Sterile Flip Seal |
| Closure Size | 13 mm |
| For Use With (Equipment) | 2 mL Serum Vial |
| Certifications/Compliance | USP 788, 85, and 71, Container Closure Integrity (CCI) certified |
Product Title
Individuate un'opportunità di miglioramento?