Learn More
Contec™ Sterile CyChlor
BIOPHARMA
Product Code. 15959875 Shop All Contec Products
Change view
Click to view available options
Volume (Metric):
1 L
5 L
Unit Size:
2 bottles
6 bottles
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Unit Size
15959875 1 L 6 bottles
15969875 5 L 2 bottles
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15959875 Supplier Contec™ Supplier No. SBC100CC
  • ONLINE EXCLUSIVE PRICE
    173.00 EUR
    141.74 EUR / Pack of 6
    Save 31.26 EUR
    18% Off
View all promotions
 Restricted Item
Estimated Shipment: 24-02-2027
Provided ready-to-use and is efficacious against bacteria and yeasts in 3 mins. Contec™ Sterile CyChlor Solution is a new broad spectrum disinfectant designed for every day use.
  • Broad spectrum efficacy in 3 mins saves time spent on biodecontamination
  • Filtered to 0.2 microns and filled in a Grade A environment ensuring the product is free from contamination and particulates
  • Suitable for Grade A and B cleanrooms
TRUSTED_SUSTAINABILITY
Specifications
Product Type Disinfectant
Form Liquid
pH 3 to 6
Sterility Sterile
Appearance Colorless
Autoclavable Not Autoclavable
Certifications/Compliance Grade A and B cleanrooms
Color Clear
Composition Fluid
Container Type Capped Container
Disposable Limited-use
Filtered Filtered
Flammable No
Quantity 6/Cs.
Volume (Metric) 1 L