Learn More
Gibco™ StemPro™ EZPassage™ Disposable Stem Cell Passaging Tool
Product Code. 11523127 Shop All Gibco Products
Change view
Click to view available options
Quantity:
10 units
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11523127 10 units Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11523127 Supplier Gibco™ Supplier No. 23181010
Curiosity Goes Beyond
Use promo code 27966
View all promotions
In Stock
Add to basket
Edge
Add to basket
Novel human embryonic stem cell passaging device that makes manual passaging of stem cell colonies more rapid and reproducible.
  • Made of a proprietary, cell culture-safe, and inert material that enables cutting of embryonic stem cell colonies growing on feeder cells or other substrates into uniform pieces for optimal passaging.
  • Outperforms existing manual and enzymatic passing methods in speed, uniformity of passaged colonies, and reliability.
  • Manually passage stem cell colonies in a fraction of the time compared with standard techniques.
  • Cut stem cell colonies into pieces of uniform size, making passaging of stem cells more reproducible.
  • Control the size for better passaging or to make embryoid bodies.
  • Cell culture-safe Passaging Tools are ready for use and packaged individually in gamma-irradiated sleeves.
  • One-time use.
  • Do not sterilize (alcohol or autoclave)
TRUSTED_SUSTAINABILITY
Specifications
Material Proprietary, Cell Culture-Safe, Inert Material
Sterility Sterile
For Use With (Application) Manual Passaging Stem Cell Colonies
Disposable Single-use
Product Type Stem Cell Passaging Tool
Cell Type Stem Cell
Product Line EZPassage, StemPro
Quantity 10 units