Learn More
STEMCELL Technologies STEMgrid™-6
Product Code. 15526110
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 Grid
Unit Size:
Pack of 5
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15526110 1 Grid Pack of 5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15526110 Supplier STEMCELL Technologies Supplier No. 27000
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Detachable grid for manual counting of hematopoietic colonies in SmartDish™ 6-well plates using an inverted microscope
  • STEMgrid™-6 is a counting grid designed to attach to the bottom of any SmartDish™ 6-well plate (Catalog #27370)
  • STEMgrid™-6 is also suitable for manual counting of hematopoietic colonies in standard 35 mm dishes, which can be placed individually onto the grid surface
  • STEMgrid™-6 contains gridlines to assist in the manual counting of hematopoietic colonies using an inverted microscope
  • The six 35 mm diameter areas of 2 x 2 mm grids precisely align with each well of a SmartDish™ 6-well plate
  • In addition, each of the six gridded areas are divided into quadrants by thicker lines, to facilitate counting of only one-quarter or one-half of the culture, and as a reference for locating colonies in different areas of a well during counting
Specifications
Quantity 1 Grid
For Use With (Application) Colony Assay