Learn More
STEMCELL Technologies STEMgrid™-6
Description
- STEMgrid™-6 is a counting grid designed to attach to the bottom of any SmartDish™ 6-well plate (Catalog #27370)
- STEMgrid™-6 is also suitable for manual counting of hematopoietic colonies in standard 35 mm dishes, which can be placed individually onto the grid surface
- STEMgrid™-6 contains gridlines to assist in the manual counting of hematopoietic colonies using an inverted microscope
- The six 35 mm diameter areas of 2 x 2 mm grids precisely align with each well of a SmartDish™ 6-well plate
- In addition, each of the six gridded areas are divided into quadrants by thicker lines, to facilitate counting of only one-quarter or one-half of the culture, and as a reference for locating colonies in different areas of a well during counting
Specifications
Specifications
| Quantity | 1 Grid |
| For Use With (Application) | Colony Assay |