Learn More
Cole-Parmer™ Integrity 10, RS-200, and RS-250 Reaction Station Accessories
Product Code. 15487854
Change view
Click to view available options
Product Type:
Reducing Sleeve
Reflux Head
Small Volume Test Tube (3 mL)
Small Volume Test Tubes (1 mL)
Unit Size:
Each
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Product Type Unit Size
15487854 Reducing Sleeve Each
15447854 Small Volume Test Tubes (1 mL) Each
15487844 Small Volume Test Tube (3 mL) Each
11806752 Reflux Head Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 15487854 Supplier Cole-Parmer™ Supplier No. 3663086
  • ONLINE EXCLUSIVE PRICE
    559.00 EUR
    434.00 EUR / Pack of 1
    Save 125.00 EUR
    22% Off
 Restricted Item
Estimated Shipment: 08-03-2027
Keep your reaction station running smoothly with these parts and accessories.
  • Suitable for use with Cole-Parmer™ Integrity 10, RS-200, or RS-250 series reaction stations
  • Choose from a wide range of accessories suitable for your reaction station including reaction vessels, probes, and software
Specifications
Product Type Reducing Sleeve
For Use With (Application) Small Volume Test Tubes (1 mL)
For Use With (Equipment) Integrity 10, RS-200, and RS-250 Reaction Stations