Learn More
R&L Enterprises™ Plated Steel Spring
An excellent option for hand use or bosshead mounting. R&L Enterprises™ Plated Steel Springs provide a gentle yet firm grip on laboratory tubing.
Brand: R&L Enterprises™ Z5904
13.06 EUR valid until 2024-03-30
Use promo code "21636" to get your promotional price.
Additional Details : Weight : 0.17100kg
Description
- With alloy rod
- For tubes up to 30mm in diameter
- Supplied in packs of 10
Specifications
Test Tube Holder Spring | |
10 Pk. | |
Steel |