missing translation for 'onlineSavingsMsg'
Learn More

Stearyl Alcohol, Sigma-Aldrich™

Product Code. 15809530
Change view
Click to view available options
Quantity:
100 g
1 kg
Unit Size:
100g
1kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15809530 100 g 100g
15806470 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15809530 Supplier Sigma Aldrich Supplier No. 8076800100
 Restricted Item
Estimated Shipment: 19-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For synthesis

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 112-92-5

Specifications

Melting Point 55°C to 60°C
Density 0.805 to 0.815 g/cm3 at 60°C
Boiling Point 330°C to 360°C/1013 hPa
MDL Number MFCD00002823
Quantity 100 g
Synonym Stearyl alcohol, Stearyl alcohol
Molecular Weight (g/mol) 270.49
Grade Research
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.