missing translation for 'onlineSavingsMsg'
Learn More

Staurosporine, MP Biomedicals™

Product Code. 11470472 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11470472 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11470472 Supplier MP Biomedicals Supplier No. 0219140080
  • ONLINE EXCLUSIVE PRICE
    204.00 EUR
    194.25 EUR / 100µg
    Save 9.75 EUR
    4% Off
 Restricted Item
Estimated Shipment: 02-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 62996-74-1
Color White
Molecular Formula C28H26N4O3
Quantity 100 μg
Synonym staurosporine, kinome_3629
InChI Key HKSZLNNOFSGOKW-ZYSRIHRCSA-N
SMILES CC12C(C(CC(O1)N3C4=CC=CC=C4C5=C6C(=C7C8=CC=CC=C8N2C7=C53)CNC6=O)NC)OC
Molecular Weight (g/mol) 466.541
PubChem CID 49831000
Formula Weight 466.5
Physical Form Powder
Chemical Name or Material Staurosporine
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.